Learn More
Andwin ScientificSupplier Diversity Partner Yamato 150L Forced Convection Oven, Economical, 220V DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner DKM610C
Economical forced convection 150L constant temperature oven with basic functions and safety features. Fixed temperature, Quick Auto stop, Auto stop, and Auto start operating modes with easy control capabilities. Settings can be made digitally with the dedicated operation menu keys or the ▲/▼ keys. Auto recovery after power failure, calibration off-set and key-lock are possible through the auxiliary functions. Operating temp ambient +10~260°C.
Catalog No. 50-189-7999