Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps Jar, Sklar, Stainless Steel. Sklar Forceps Jars are ideal for holding multiple pairs of forceps and other instruments. This jar is measures 2-1/4 in. x 7 in. with a 3-11/16 in. base. They are sold in packs of 12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 10154512
Forceps Jar, Sklar, Stainless Steel. Sklar Forceps Jars are ideal for holding multiple pairs of forceps and other instruments upright so they can be readily accessed. The cylandrical shape and uncovered mouth allow handles to rest where they can be easily grasped. The base, of a larger circumference than the rest of the container, ensures the jar does not tip. This jar is measures 2-1/4 in. x 7 in. with a 3-11/16 in. base. They are sold in packs of 12.
Catalog No. 50-143-7056