Learn More
Innovating Science™ Forensic Chemistry of Blood Types Kit

Classify blood based on the presence or absence of erythrocytes on the surface of red blood cells.
$27.50 - $65.25
Specifications
Certifications/Compliance | 49 CFR 173.4 |
---|---|
Color | White |
Grade | 7 to 14 |
For Use With (Application) | For forensic chemistry of blood typing |
Quantity | 1 |
Includes: Teacher's manual, student guide, analysis copy-masters, blood typing trays, four simulated blood samples (victim, suspect #1, suspect #2 and crime scene), ABO/Rh blood typing anti-sera, mixing sticks. Enough materials for 15 tests.
Catalog Number | Mfr. No. | Includes | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Product Type | Price | Quantity & Availability | ||||
S96644R
![]() |
Innovating Science™
IS9008REF |
Simulated blood samples, crime scene, abo/rh blood typing, anti-sera, Teacher's manual and student study guide copymasters | Forensic Chemistry of Blood Types Refill |
Each for $27.50
|
|
||||
S96644
![]() |
Innovating Science™
IS9008 |
Blood typing trays, 4 simulated blood samples, crime scene, 1 set abo/rh blood typing anti-sera, 1 pkg mixing stick, Teacher's manual and student study guide copymasters | Forensic Chemistry of Blood Types Kit |
Each for $65.25
|
|
||||
Description
Blood type, an inherited characteristic, is valuable to know in that it affects medical procedures, such as surgery and transfusions, paternity testing, as well as serving as evidence in criminal investigations.
Uses Innovating Science Simulated Blood; safe for classroom use.
Specifications
49 CFR 173.4 | |
7 to 14 | |
1 |
White | |
For forensic chemistry of blood typing | |
15 Lab Groups |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.