Learn More
Formlabs Inc FORM 3 RESIN TANK V2
Supplier:  Formlabs Inc RTF302
Encompass
Form 3 Resin Tank V2Low Force Stereolithography (LFS) 3D printing uses linear illumination and a flexible film tank to turn liquid resin into flawless parts. A key component of the LFS process, the strong dual-layer film at the bottom of the Form 3 Resin Tank allows for a dynamic and precise print process.For use with Form 3 and Form 3B.
Catalog No. NC1820353
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product