Learn More
Thermo Scientific™ Forma™ Forma™ FDE Series -40°C Ultra-Low Temperature Freezer Package with Racks, Boxes, and Chart Recorder Available on GSA/VA Contract for Federal Government customers only.
Forma™ FDE -40°C ultra-low temperature freezer package includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed chart recorder
$16803.00 - $17892.00
Specifications
Temperature Range -10°C to -40°C
View More Specs
Products 4
Catalog Number Mfr. No. Electrical Requirements Certifications/Compliance Price Quantity & Availability  
Catalog Number Mfr. No. Electrical Requirements Certifications/Compliance Price Quantity & Availability  
FDE540FARCR Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™ Forma™
FDE50040FARCR
115V, 60Hz cULus
Each for $16,803.00
Add to cart
FDE540FDRCR Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™ Forma™
FDE50040FDRCR
208 - 230V, 60Hz cULus
Each for $16,803.00
Add to cart
FDE640FARCR Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™ Forma™
FDE60040FARCR
115V, 60Hz cULus
Each for $17,892.00
Add to cart
FDE640FDRCR Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™ Forma™
FDE60040FDRCR
208 - 230V, 60Hz cULus
Each for $17,892.00
Add to cart
Description
Description
Forma™ FDE -40°C ultra-low temperature freezer packages include one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed chart recorder.
Package includes:
  • One Thermo Scientific Forma FDE -40°C Ultra-Low temperature freezer
  • 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
  • Factory-installed chart recorder
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
-10°C to -40°C