Learn More
Thermo Scientific™ Forma™ Forma™ FDE Series -40°C Ultra-Low Temperature Freezer Package with Racks, Boxes, and CO2 Back-up System
GSA_VA
Forma™ FDE -40°C ultra-low temperature freezer package includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed CO2 back-up system
$20415.00 - $23382.00
Specifications
Temperature Range -10°C to -40°C
Door Style 1 Outer, 4 Inner
No. of Doors 5
Insulation Water-Blown Polyurethane Foam
Interior Stainless Steel
View More Specs
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Products 6
Catalog Number Mfr. No. Electrical Requirements Certifications/Compliance Holds Price Quantity & Availability  
Catalog Number Mfr. No. Electrical Requirements Certifications/Compliance Holds Price Quantity & Availability  
FDE440FARCO
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE40040FARCO
115V, 60Hz cULus Boxes (400)
Each for $20,415.00
Add to cart
FDE440FDRCO
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE40040FDRCO
208 - 230V, 60Hz cULus Boxes (400)
Each for $20,415.00
Add to cart
FDE540FARCO
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE50040FARCO
115V, 60Hz cULus Boxes (500)
Each for $22,012.00
Add to cart
FDE540FDRCO
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE50040FDRCO
208 - 230V, 60Hz cULus Boxes (500)
Each for $22,012.00
Add to cart
FDE640FARCO
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE60040FARCO
115V, 60Hz cULus Boxes (600)
Each for $23,382.00
Add to cart
FDE640FDRCO
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE60040FDRCO
208 - 230V, 60Hz cULus Boxes (600)
Each for $23,382.00
Add to cart
Description
Description
Forma™ FDE -40°C ultra-low temperature freezer packages include one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed CO2 back-up system.
Package includes:
  • One Thermo Scientific Forma FDE -40°C Ultra-Low temperature freezer
  • 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
  • Factory-installed CO2 back-up system (CO2 tank not included)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
-10°C to -40°C
5
Stainless Steel
Upright
H-Drive Compressor
1 Outer, 4 Inner
Water-Blown Polyurethane Foam
2
Alarm, Recorder