Learn More
Thermo Scientific™ Forma™ Forma™ FDE Series -40°C Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System
GSA_VA
Forma™ FDE -40°C ultra-low temperature freezer package includes one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed LN2 back-up system
$20415.00 - $23382.00
Specifications
Temperature Range -10°C to -40°C
Door Style 1 Outer, 4 Inner
No. of Doors 5
Insulation Water-Blown Polyurethane Foam
Interior Stainless Steel
View More Specs
Products 6
Catalog Number Mfr. No. Electrical Requirements Certifications/Compliance Includes Price Quantity & Availability  
Catalog Number Mfr. No. Electrical Requirements Certifications/Compliance Includes Price Quantity & Availability  
FDE440FARLN
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE40040FARLN
115V, 60Hz cULus 400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and a factory installed LN2 backup system
Each for $20,415.00
Add to cart
FDE440FDRLN
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE40040FDRLN
208 - 230V, 60Hz cULus 400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and a factory installed LN2 backup system
Each for $20,415.00
Add to cart
FDE540FARLN
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE50040FARLN
115V, 60Hz cULus 500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and a factory installed LN2 backup system
Each for $22,012.00
Add to cart
FDE540FDRLN
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE50040FDRLN
208 - 230V, 60Hz cULus 500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and a factory installed LN2 backup system
Each for $22,012.00
Add to cart
FDE640FARLN
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE60040FARLN
115V, 60Hz cULus 600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and a factory installed LN2 backup system
Each for $23,365.00
Add to cart
FDE640FDRLN
GSA_VA
View Documents
Thermo Scientific™ Forma™
FDE60040FDRLN
208 - 230V, 60Hz cULus 600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and a factory installed LN2 backup system
Each for $23,382.00
Add to cart
Description
Description
Forma™ FDE -40°C ultra-low temperature freezer packages include one ULT freezer, two shelves of sliding drawer racks and boxes, and a factory-installed LN2 back-up system.
Package includes:
  • One Thermo Scientific Forma FDE -40°C Ultra-Low temperature freezer
  • 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
  • Factory-installed LN2 back-up system (LN2 tank not included)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
-10°C to -40°C
5
Stainless Steel
Upright
H-Drive Compressor
1 Outer, 4 Inner
Water-Blown Polyurethane Foam
2
Alarm, Recorder