Learn More
Thermo Scientific™ Forma™ FDE Series Ultra-Low Temperature Freezer Package with Racks and Boxes
GSA_VA
Forma™ FDE ultra-low temperature freezer package includes one ULT freezer and two shelves of sliding drawer racks and boxes
$19010.00 - $30850.00
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Includes: 300 box capacity freezer, 6 sliding drawer racks, 150 2-inch boxes with 100-count cell dividers
Products 8
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Includes Certifications/Compliance Electrical Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W x H) Exterior Includes Certifications/Compliance Electrical Requirements Price Quantity & Availability  
FDE386FARK
GSA_VA
View Documents
Thermo Scientific™
FDE30086FARK
38.5 x 23 x 78 in. (97.7 x 58.4 x 198.1 cm) 300 box capacity freezer, 6 sliding drawer racks, 150 2-inch boxes with 100-count cell dividers cULus 115 V 60 Hz
Each for $19,010.00
Add to cart
FDE386FDRK
GSA_VA
Accessories View Documents
Thermo Scientific™
FDE30086FDRK
38.5 x 23 x 78 in. (97.7 x 58.4 x 198.1 cm) 300 box capacity freezer, 6 sliding drawer racks, 150 2-inch boxes with 100-count cell dividers cULus 208 - 230 V 60 Hz
Each for $19,010.00
Add to cart
FDE486FARK
GSA_VA
View Documents
Thermo Scientific™
FDE40086FARK
38.5 x 28.3 x 78 in. (97.7 x 71.9 x 198.1 cm) 400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers cULus 115 V 60 Hz
Each for $21,980.00
Add to cart
FDE486FDRK
GSA_VA
View Documents
Thermo Scientific™
FDE40086FDRK
38.5 x 28.3 x 78 in. (97.7 x 71.9 x 198.1 cm) 400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers cULus 208 - 230 V 60 Hz N/A N/A
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance.
FDE586FARK
GSA_VA
View Documents
Thermo Scientific™
FDE50086FARK
38.5 x 34.0 x 78 in. (97.7 x 86.4 x 198.1 cm) 500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers cULus 115 V 60 Hz
Each for $25,550.00
Add to cart
FDE586FDRK
GSA_VA
Accessories View Documents
Thermo Scientific™
FDE50086FDRK
38.5 x 34.0 x 78 in. (97.7 x 86.4 x 198.1 cm) 500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers cULus 208 - 230 V 60 Hz
Each for $25,550.00
Add to cart
FDE686FARK
GSA_VA
View Documents
Thermo Scientific™
FDE60086FARK
38.5 x 39.6 x 78 in. (97.7 x 100.6 x 198.1 cm) 600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers cULus 115 V 60 Hz
Each for $30,850.00
Add to cart
FDE686FDRK
GSA_VA
View Documents
Thermo Scientific™
FDE60086FDRK
38.5 x 39.6 x 78 in. (97.7 x 100.6 x 198.1 cm) 600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers cULus 208 - 230 V 60 Hz N/A N/A
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance.
Description
Description
Part of the Standard Performance (STP) family, Thermo Scientific™ Forma™ FDE ultra-low temperature freezer packages include one ULT freezer and two shelves of sliding drawer racks and boxes.
Package includes:
  • One Thermo Scientific Forma FDE Ultra-Low temperature freezer
  • 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.