Learn More
Thermo Scientific™ Forma™ Steri-cycle™ i160 CO2 Incubator with Cell Locker™ System
Protected chambers for your most sensitive cells.  A breakthrough in cell culture management that maximizes your sample security.
Manufacturer:  Thermo Scientific™ 51032172
 View more versions of this product
Catalog No. 11-675-964
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The Thermo Scientific™ Forma™ Steri-cycle™ i160 CO2 incubator with the Thermo Scientific™ Cell Locker™ system is an innovative solution of removable, protected chambers designed for improved culturing efficiency and security for sensitive cultures. Six individual, autoclavable polycarbonate chambers divide the incubator chamber, isolating individual cell types or projects. Each Cell Locker chamber has dual 0.2µm membrane filters that permit air circulation but exclude microbial contaminants.When a Cell Locker chamber is opened, the others remain stable, preserving the in vitro environment.
Order Info
Please order Cell Lockers chambers separately if not included. 50151650 is a single Cell Locker chamber with membrane filters with stainless steel work tray and transport cover.; 50154739 is a single Cell Locker chamber with membrane filters with copper work tray and transport cover
Specifications
Specifications
Electropolished stainless steel
IR180SI
Covered integrated water reservoir
Ambient +3° to 55°C
CE, CSA
5.8 cu.ft., 165L per chamber
34.6 x 25.1 x 35.4 in. (88 x 63.7 x 90cm)
183 lb, 83 kg
1 to 20%
Optional 1-21% or 5-90%
≥93% @ 37°C
USB, and optional 4-20mA
22.7 x 18.5 x 23.9 in. (57.6 x 47 x 60.7cm)
120V 50/60Hz
Single 165L stainless steel chamber configured for Cell Locker system, (Cell Locker chambers ordered separately)