Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 10% (v/v) Buffered Solution, Spectrum™

Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
50-00-0,7732-18-5,67-56-1
50-00-0,7732-18-5,67-56-1
Packaging Cubitainer
Quantity 20 L
pH 6.9 to 7.1
Appearance Clear, Colorless
Concentration or Composition (by Analyte or Components) 38 - 42 g/L
Grade Chemical Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.