Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 10% (v/v) Neutralized and Buffered Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
50-00-0,7732-18-5,7558-79-4
50-00-0,7732-18-5,7558-79-4
Packaging Poly Bottle
Quantity 4 L
pH 6.8 to 7.2
Grade Chemical Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.