Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 37% by Weight (With Preservative/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1L
500mL
20L
4L
200L
Packaging:
Glass Bottle
Poly Bottle
Poly Drum
Poly Pail
PolyPac™
50-00-0,67-56-1,7732-18-5
CH2O
30.026
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde
712
CHEBI:16842
formaldehyde
C=O
50-00-0,67-56-1,7732-18-5
CH2O
30.026
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde
712
CHEBI:16842
formaldehyde
C=O

Stabilized with 10-15% methanol

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Melting Point 0°C
Ignition Residue 0.005% max.
Methanol 9 to 15% v/v
Chloride 5ppm max.
Boiling Point 101°C
Iron (Fe) 2ppm max.
Packaging Glass Bottle
Heavy Metals (as Pb) 5ppm max.
Sulfate 0.002% max.
Quantity 4L
Color Colorless
Titratable Acid 0.006mEq/g max.
Assay Percent Range 36.5 to 38.0 %
Linear Formula HCHO
Concentration or Composition (by Analyte or Components) Stabilized with 10-15% Methanol
Formula Weight 30.03
Physical Form Liquid
Percent Purity 36.5 to 38.0%
Grade Certified ACS
Chemical Name or Material Formaldehyde, 37% by Weight, With Preservative
Show More Show Less

DANGER!

Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid