Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 37% Solution, BAKER ANALYZED™ ACS Reagent, J.T.Baker™

Click to view available options
Quantity:
19L
150mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail

Chemical Identifiers

50-00-0
CH2O
30.026
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
CHEBI:16842
formaldehyde
C=O
Packaging Poly Pail
Flash Point 60°C
Quantity 19L
Formula Weight 30.03
Chemical Name or Material Formaldehyde
Assay 36.5 to 38.0%
Grade ACS Grade