Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formaldehyde, 37% Solution, BAKER ANALYZED™ ACS Reagent, J.T.Baker™
Click to view available options
Quantity:
19L
150mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
Specifications
Flash Point | 60°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 150mL |
Formula Weight | 30.03 |
Percent Purity | 36.5 to 38.0% |
Grade | ACS |
Chemical Name or Material | Formaldehyde |