Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Plastic bottle
Plastic drum
50-00-0,7732-18-5,67-56-1
CH2O
30.026
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde
712
CHEBI:16842
formaldehyde
C=O
50-00-0,7732-18-5,67-56-1
CH2O
30.026
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde
712
CHEBI:16842
formaldehyde
C=O
Refractive Index 1.3755 to 1.3775
Flash Point 50°C
Packaging Plastic drum
Boiling Point 97.0°C
Density 1.0830g/mL
Melting Point -15.0°C
Quantity 25L
Additional Information Vapor Pressure: 2mbar at 20°C
Free Acid 0.1% max.
Stabilizer 5 to 15% methanol
Color Clear
Linear Formula HCHO
Fieser 01,397; 02,200; 04,238; 05,312; 06,264; 07,158; 08,231; 09,224; 10,186; 11,240; 12,232; 14,168; 15,161; 16,171
Merck Index 15, 4263
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and acetone
Formula Weight 30.03
Physical Form Solution
Percent Purity 35.0 to 39.0%
Chemical Name or Material Formaldehyde, 37 wt% Solution in Water, Stabilized with 10 to 15% Methanol
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage. May cause an allergic skin reaction. Causes damage to organs. Toxic in contact with skin. Toxic if inhaled. Toxic if swallowed. May cause cancer. Flammable liquid and vapor. Suspected of causing genetic defects if inhaled. May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Do not handle until all safety precautions have been read and understood.

Danger