Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Formaldehyde, 4% in PBS

Catalog No. p-7076543
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60401AK 250 mL
AAJ60401AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60401AK Supplier Thermo Scientific Chemicals Supplier No. J60401AK
Add to Cart

Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses. | CAS: 50-00-0 | CH2O | 30.03 g/mol

Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Packaging Plastic bottle
Quantity 250 mL
Solubility Information Soluble in water,ethanol,acetone,and ether.
Chemical Name or Material Formaldehyde
Hazard Category H315-H317-H319-H341-H350
Precautionary Statement P201-P202-P261-P264b-P272-P280-P281-P302+P352-P305+P351+P338-P308+P313-P333+P313-P362-P501c
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.