Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, Para (Laboratory), Fisher Chemical

Catalog Number t353500
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
30525-89-4, 50-00-0
CH2O
30.026
MFCD00133991
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
712
CHEBI:16842
formaldehyde
C=O
30525-89-4, 50-00-0
CH2O
30.026
MFCD00133991
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
712
CHEBI:16842
formaldehyde
C=O
Melting Point 120°C
Color White
Density 1.46g/cm³
pH 3.5 to 5.0
Packaging Poly Bottle
Quantity 500 g
UN Number 2213
Vapor Pressure 1.2mmHg at 25°C
Grade Laboratory
Physical Form Solid
Chemical Name or Material Formaldehyde, Para
Show More Show Less
Hazard Category Signal Word: DANGER!
Precautionary Statement Emergency Overview
Flammable solid. Harmful by inhalation and if swallowed. Irritating to eyes, respiratory system and skin. Possible cancer hazard. May cause cancer based on animal data. May cause pulmonary edema.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit