Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde Sodium Bisulfite Addition Compound 95%, ACROS Organics™

Click to view available options
Quantity:
5g
1kg
5kg
Packaging:
Glass Bottle
Plastic bottle
870-72-4
CH3NaO4S
134.081
MFCD00044664
UOULCEYHQNCFFH-UHFFFAOYSA-M
sodium hydroxymethanesulfonate,formaldehyde sodium bisulfite,formbis,sodium formaldehyde bisulfite,sodium hydroxymethanesulphonate,methanesulfonic acid, hydroxy-, monosodium salt,sodium hydroxymethane sulfonate,sodium formaldehydebisulfite,unii-i566kha05f,methylolsulfonic acid sodium salt
2723822
sodium;hydroxymethanesulfonate
C(O)S(=O)(=O)[O-].[Na+]
870-72-4
CH3NaO4S
134.081
MFCD00044664
UOULCEYHQNCFFH-UHFFFAOYSA-M
sodium hydroxymethanesulfonate,formaldehyde sodium bisulfite,formbis,sodium formaldehyde bisulfite,sodium hydroxymethanesulphonate,methanesulfonic acid, hydroxy-, monosodium salt,sodium hydroxymethane sulfonate,sodium formaldehydebisulfite,unii-i566kha05f,methylolsulfonic acid sodium salt
2723822
sodium;hydroxymethanesulfonate
C(O)S(=O)(=O)[O-].[Na+]
Quantity 1kg
Color White
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 95%
Linear Formula HOCH2SO3Na
Formula Weight 134.08
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material Formaldehyde sodium bisulfite addition compound, 95%

HYGROSCOPIC

EINECSNumber : 212-800-9

RTECSNumber : PB2200000

TSCA : TSCA