Alfa Aesar™ Formaldoxime trimer hydrochloride, 95%

Manufacturer: Alfa Aesar™ B2506322

Quantity 100g

 View more versions of this product

Catalog No. AAB2506322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6286-29-9
    Molecular Formula C3H9N3O3·HCl
    Formula Weight 171.57
    MDL Number MFCD00012778
    UN Number UN2811
    Beilstein 3690722
    Synonym Formaldehyde oxime trimer hydrochloride; Hexahydro-1,3,5-trihydroxy-1,3,5-triazine hydrochloride
    Chemical Name or Material Formaldoxime trimer hydrochloride

    Specifications

    Formula Weight 171.57
    Melting Point ≈128°C (dec.)
    Quantity 100g
    Molecular Formula C3H9N3O3·HCl
    MDL Number MFCD00012778
    CAS 6286-29-9
    UN Number UN2811
    Beilstein 3690722
    Synonym Formaldehyde oxime trimer hydrochloride; Hexahydro-1,3,5-trihydroxy-1,3,5-triazine hydrochloride
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    RTECS Number TQ0300000
    Health Hazard 2 GHS H Statement
    H300-H310-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material Formaldoxime trimer hydrochloride
    Assay 95%

    GHS H Statement
    H300-H310-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit