Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Formaldoxime trimer hydrochloride, 95%

Click to view available options
Quantity:
10g
50g
100g
6286-29-9
C3H10ClN3O3
171.581
MFCD00012778
VRHJXINSVWQXEB-UHFFFAOYSA-N
formaldoxime trimer hydrochloride,sym-triazinetriol hydrochloride,triformoxime hydrochloride,1,3,5-triazinane-1,3,5-triol hydrochloride,1,3,5-triazaperhydroine-1,3,5-triol, chloride,1,3,5-triazine, hexahydro-1,3,5-trihydroxy-, hydrochloride
15919123
1,3,5-trihydroxy-1,3,5-triazinane;hydrochloride
C1N(CN(CN1O)O)O.Cl
6286-29-9
C3H10ClN3O3
171.581
MFCD00012778
VRHJXINSVWQXEB-UHFFFAOYSA-N
formaldoxime trimer hydrochloride,sym-triazinetriol hydrochloride,triformoxime hydrochloride,1,3,5-triazinane-1,3,5-triol hydrochloride,1,3,5-triazaperhydroine-1,3,5-triol, chloride,1,3,5-triazine, hexahydro-1,3,5-trihydroxy-, hydrochloride
15919123
1,3,5-trihydroxy-1,3,5-triazinane;hydrochloride
C1N(CN(CN1O)O)O.Cl
Sensitivity Hygroscopic
Melting Point ~128°C (decomposition)
Quantity 10g
UN Number UN2811
Beilstein 3690722
Formula Weight 171.57
Percent Purity 95%
Chemical Name or Material Formaldoxime trimer hydrochloride

GHS H Statement
H300-H310-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H300+H310-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 222-440-4

RTECSNumber : TQ0300000

TSCA : No

Recommended Storage : Ambient temperatures