Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formalin-Aceto-Alcohol (F-A-A Mixture), for Rapid Fixation of Tissue, Ricca Chemical
Rapid Fixation of Tissue
Specifications
Chemical Name or Material | Formalin-Aceto-Alcohol |
---|---|
Physical Form | Liquid |
Name Note | F-A-A Mixture |
CAS Min % | 43.0, 34.92, 17.29, 2.82 |
CAS Max % | 45.0, 36.34, 18.0, 2.94 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
3210-16
|
Ricca Chemical Company
321016 |
500 mL | Natural Poly Bottle |
Each for $105.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
3210-32
|
Ricca Chemical Company
321032 |
1 L | Natural Poly Bottle |
Each for $168.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Specifications
Formalin-Aceto-Alcohol | |
F-A-A Mixture | |
45.0, 36.34, 18.0, 2.94 | |
Natural Poly Bottle |
Liquid | |
43.0, 34.92, 17.29, 2.82 | |
7732-18-5, 50-00-0, 64-19-7, 7732-18-5 | |
Colorless |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title