Learn More
Carolina™ Formalin Bullfrogs
For use in introducing students to vertebrate anatomy. Carolina™ Formalin Bullfrogs are ideal for studying amphibian anatomy, adaptations, and body systems.
Supplier: Carolina™ S227102
This item is not returnable.
View return policy
Description
- Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
- Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
- INJECTIONS: Plain: no color injection; Double: red arteries, blue hepatic
Specifications
Preserved Specimen | |
Plain | |
12.7+ cm | |
Bulk Bag |
Formalin | |
5+ in. | |
5/Bag |