Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Formalin Cats
For use in advanced biology, anatomy and physiology classes. Carolina™ Formalin Cats are ideal dissection specimens.
Supplier: Carolina™ 228080
Description
- Felis catus
- Similar to human anatomy
- Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
- Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
- INJECTIONS: Plain: n
Specifications
Preserved Specimen | |
Triple | |
35.5 to 45.7 cm | |
Bag |
Formalin | |
14 to 18 in. | |
1/Bag |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction