Learn More
Carolina™ Formalin Dogfish Sharks
For use in exploring anatomy of cartilaginous fish. Carolina™ Formalin Dogfish Sharks are ideal for comparative and vertebrate anatomy classes.
Supplier: Carolina™ 226741
Description
- Member of class Chondrichthyes
- Mature specimens
- Characteristics: cartilaginous skeleton, five pairs of gill slits, no swim bladder or operculum
- Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
Specifications
Preserved Specimen | |
Double | |
68.5+ cm | |
Bag |
Formalin | |
27+ in. | |
1/Bag |