Learn More
Carolina™ Formalin Earthworms
For use in introducing students to invertebrate anatomy. Carolina™ Formalin Earthworms are ideal introductory dissection specimens.
Supplier: Carolina™ 225005
This item is not returnable.
View return policy
Description
- Lumbricus terrestris
- Phylum Annelida, segmented worms
- Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
- Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
- LAT
Specifications
Preserved Specimen | |
Injected | |
22.8 to 30.4 cm | |
Tube |
Formalin | |
9 to 12 in. | |
10/Tube |