Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, ≥99.0% (GC), Reagent Grade, Honeywell™

Click to view available options
Quantity:
5mL
250mL
500mL
1L
2.5L
4L
Packaging:
Glass bottle
Poly bottle
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O

Reagent Grade,≥99.0% (GC)

  • ≥95% for general laboratory use.
Linear Formula HCONH2
UN Number NONH for all modes of transport
Beilstein 505995
Formula Weight 45.04g/mol
Percent Purity ≥99.0% (GC)
Density 1.134g/mL at 25°C
Ignition Point 932°F
Odor Weak
Packaging Glass bottle
Solubility in Water Completely miscible
Vapor Pressure 0.08mmHg (20°C), 30mmHg (129°C)
Vapor Density 1.55 (vs air)
Flash Point 175°C (347°F)
Physical State Liquid
Boiling Point 210°C (1013hPa)
Color Colorless
Melting Point 1°C to 3°C
pH 4.0 to 5.0 (at 20°C)
Quantity 5mL
Chemical Name or Material Formamide
Show More Show Less

P201-P260-P280-P308 + P313

RTECSNumber : 200-842-0