Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, 99%, Thermo Scientific™

Click to view available options
Quantity:
500g
2500g
10kg
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
Applications
Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabiliser in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assemb
Linear Formula HCONH2
Beilstein 505995
Merck Index 14,4237
Solubility Information It is miscible with water.
Formula Weight 45.04
Percent Purity 99%
Density 1.134
Odor Ammonia-like
Refractive Index 1.4475
Flash Point 150°C (302°F)
Boiling Point 210°C
Melting Point 2°C
Quantity 500g
Sensitivity Hygroscopic
Chemical Name or Material Formamide
Show More Show Less

GHS H Statement
H360;
May damage fertility or the unborn child.

P201-P202-P260-P281-P308+P313-P501c

H351-H360FD-H373

EINECSNumber : 200-842-0

RTECSNumber : LQ0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only