Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, 99.5+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Plastic drum
75-12-7
CH3NO
45.041
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88
713
CHEBI:48431
formamide
C(=O)N
75-12-7
CH3NO
45.041
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88
713
CHEBI:48431
formamide
C(=O)N
Assay Percent Range 99.5+%
Linear Formula HCONH2
Beilstein 02, 26
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
Formula Weight 45.04
Physical Form Liquid
Percent Purity ≥99.5%
Grade Extra Pure
Density 1.1330g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4460 to 1.4480
Residue after Evaporation 0.05% max.
Solubility Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
Specific Gravity 1.133
Water 0.1% max. (Coulometric)
Flash Point 175°C
Viscosity 3.75 mPa.s (20°C)
Boiling Point 210.0°C
Color Colorless to Yellow
Melting Point 2.0°C to 3.0°C
Quantity 2.5L
Chemical Name or Material Formamide, 99.5+%
Show More Show Less

GHS P Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. Suspected of causing cancer if swallowed.

GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required.

Danger

EINECSNumber : 200-842-0

RTECSNumber : LQ0525000

TSCA : TSCA