Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, 99.5% min., ACS, MilliporeSigma™

Click to view available options
Quantity:
500g
1kg
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
Assay Percent Range 99.5% min.
Linear Formula HCONH2
Formula Weight 45.4
Physical Form Slightly Viscous Liquid
Grade ACS/EP/Reagent
Density 1.13g/cm3 (20°C)
Explosion Limit 2.7 to 19.0%(V)
Ignition Point 500°C
Vapor Pressure 0.08 hPa (20°C)
Color Colorless
Quantity 500g
Chemical Name or Material Formamide, For ACS analysis
Show More Show Less

Recommended Storage : Store at +15°C to +25°C