Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, 99%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
For molecular biology
Absorbance (0.5 M in water), (0.5 M in water) at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.5M in water)
Assay Percent Range 99%
Linear Formula HCONH2
Beilstein 02, 26
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
Formula Weight 45.04
Physical Form Liquid
Percent Purity 99%
Grade Molecular Biology
Copper (Cu) 0.1ppm max.
Density 1.1330g/mL
DNase RNase Protease act none detected
Iron (Fe) 0.5ppm max.
Lead (Pb) 0.5ppm max.
Packaging Glass Bottle
Refractive Index 1.4460 to 1.4480
Solubility Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
Specific Gravity 1.133
Trace Analysis 0.5ppm max.
Flash Point 175°C
Viscosity 3.75 mPa.s (20°C)
Boiling Point 210.0°C
Color Undesignated
Melting Point 2.0°C to 3.0°C
pH 7 to 9 (1M in 0.1M KCl)
Quantity 500mL
Chemical Name or Material Formamide, DNase, RNase and Protease Free
Show More Show Less

GHS P Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. Suspected of causing cancer if swallowed.

GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required.

Danger

EINECSNumber : 200-842-0

RTECSNumber : LQ0525000

TSCA : TSCA

RUO – Research Use Only