Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, ≥99%, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
75-12-7
CH3NO
45.041
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
C(=O)N
75-12-7
CH3NO
45.041
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
C(=O)N
  • For use in nucleic acid hybridization buffers, specialized sequencing gels and other molecular biology applications
  • Used to reduce Tm of nucleic acid hybrids
  • Used to achieve stronger denaturing conditions in sequencing gel along wi
Linear Formula HCONH2
Beilstein 505995
Formula Weight 45.04
Physical Form Liquid
Percent Purity ≥99%
Grade Ultrapure
Density 1.134
Packaging Plastic bottle
Refractive Index 1.447
Specific Gravity 1.134
Flash Point 154°C
Boiling Point 210°C
Melting Point 2°C
Quantity 1 kg
Chemical Name or Material Formamide
Show More Show Less

P201-P202-P260-P281-P308+P313-P501c

H351-H360FD-H373

EINECSNumber : 200-842-0

RTECSNumber : LQ0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only