Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
53 gal
Packaging:
Amber Glass Bottle
Drum
Poly Drum
75-12-7
75-12-7
CAS Max % 1
Percent Purity 0.995
Grade ACS
Packaging Amber Glass Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.