Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, Honeywell Riedel-de Haen™

Click to view available options
2 L

Spectrophotometric Grade, ≥99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
CAS 75-12-7
PubChem CID 713
ChEBI CHEBI:48431
Physical Form Liquid
Percent Purity min. 99.0 %
Grade Spectroscopy Solvents
Packaging Glass Bottle
Flash Point 175 °C
Quantity 2 L

Recommended Storage : Room Temperature