Learn More
Seracare Life Sciences IncSupplier Diversity Partner FORMAMIDE HYBRIDIZATION BUFFER  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 59600023
Formamide Hybridization Buffer is a formamide-based solution for use in the prehybridization and hybridization of biotinylated nucleic acid probes to nucleic acids fixed on membranes. As a prehybridization solution, the buffer blocks sites on the membrane and prevents nonspecific binding of biotinylated probe. As a hybridization solution, the buffer facilitates binding of biotinylated probes to homologous RNA or DNA on a membrane.
Catalog No. 50-674-49