Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Formamide, Molecular Biology Grade

Click to view available options
Quantity:
100mL
500mL
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Beilstein 505995
Merck Index 14,4237
Formula Weight 45.04
Physical Form Liquid
Grade Molecular Biology
Density 1.134 g/mL
Odor Ammonia-like
Refractive Index 1.4475
Flash Point 150° (302°F)
Boiling Point 210°C
Color Colorless
Melting Point 2°C
Quantity 100mL
Sensitivity Hygroscopic
Chemical Name or Material Formamide
Show More Show Less

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P260-P281-P308+P313-P501c

H351-H360FD-H373

EINECSNumber : 200-842-0

RTECSNumber : LQ0525000

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

RUO – Research Use Only