Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Formamide, Ultra Pure, 99.5% min

Click to view available options
Quantity:
100mL
250mL
75-12-7
CH3NO
45.041
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88
713
CHEBI:48431
formamide
C(=O)N
75-12-7
CH3NO
45.041
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide,carbamaldehyde,formimidic acid,formic acid, amide,formamid,methanoic acid, amide,amid kyseliny mravenci,amid kyseliny mravenci czech,formic amide,hsdb 88
713
CHEBI:48431
formamide
C(=O)N
Applications
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.

Solubility
Soluble in water (20 mg/ml ), methanol, ethanol and acetone. Slightly soluble in ether.

Notes
Hygroscopic. Store at room temperature. Store away from water or moisture.
Beilstein 505995
Formula Weight 45.04
Physical Form Liquid
Percent Purity ≥99.5%
Density 1.134g/mL
Odor Ammonia-like
Refractive Index 1.4475
Boiling Point 210°C
Color Colorless
Melting Point 2°C
Quantity 100mL
Sensitivity Hygroscopic
Chemical Name or Material Formamide, Ultra Pure
Show More Show Less

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : LQ0525000

TSCA : TSCA