Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

75-12-7
CH3NO
45.041
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
CHEBI:48431

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For denaturing nucleic acids prior to analysis
CAS 75-12-7
Molecular Formula CH3NO
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
SMILES C(=O)N
IUPAC Name formamide
Molecular Weight (g/mol) 45.041
PubChem CID 713
ChEBI CHEBI:48431
Formula Weight 45.04
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.13kg
Flash Point 154.00°C
Quantity 100mL
Chemical Name or Material Formamide
Grade Ultrapure Bioreagent
Show More Show Less