Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamidine acetate, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
3473-63-0
CH4N2·C2H4O2
104.11
MFCD00012866
XPOLVIIHTDKJRY-UHFFFAOYSA-N
formamidine acetate,methanimidamide, monoacetate,formamidinium acetate,acetic acid; methanimidamide,formamidine acetate salt,formamidine, monoacetate,formimidamide acetate,imidoformamide acetate,acetic acid; formamidine,methanimidamide monoacetate
160693
acetic acid;methanimidamide
CC(=O)O.C(=N)N
3473-63-0
CH4N2·C2H4O2
104.11
MFCD00012866
XPOLVIIHTDKJRY-UHFFFAOYSA-N
formamidine acetate,methanimidamide, monoacetate,formamidinium acetate,acetic acid; methanimidamide,formamidine acetate salt,formamidine, monoacetate,formimidamide acetate,imidoformamide acetate,acetic acid; formamidine,methanimidamide monoacetate
160693
acetic acid;methanimidamide
CC(=O)O.C(=N)N
Melting Point 157°C to 161°C
Specific Gravity 1.124
Density 1.1240g/mL
Packaging Plastic bottle
Color Colorless to Yellow
Quantity 500g
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula HC(=NH)NH2·CH3CO2H
Fieser 01,403; 02,202; 03,147
Solubility Information Solubility in water: 832g/L (21°C)
Formula Weight 104.11
Physical Form Liquid
Percent Purity ≥98.5% (Total base)
Chemical Name or Material Formamidine acetate, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 222-442-5

TSCA : TSCA