Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamidinesulfinic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
1758-73-2
CH4N2O2S
108.11
MFCD00002397
FYOWZTWVYZOZSI-UHFFFAOYSA-N
thiourea dioxide,formamidinesulfinic acid,amino imino methanesulfinic acid,formamidine sulfinic acid,manofast,thiourea s,s-dioxide,aimsa,methanesulfinic acid, aminoimino,carbamimidoylsulfinic acid,methenesulfinic acid, aminoimino
61274
amino(imino)methanesulfinic acid
C(=N)(N)S(=O)O
1758-73-2
CH4N2O2S
108.11
MFCD00002397
FYOWZTWVYZOZSI-UHFFFAOYSA-N
thiourea dioxide,formamidinesulfinic acid,amino imino methanesulfinic acid,formamidine sulfinic acid,manofast,thiourea s,s-dioxide,aimsa,methanesulfinic acid, aminoimino,carbamimidoylsulfinic acid,methenesulfinic acid, aminoimino
61274
amino(imino)methanesulfinic acid
C(=N)(N)S(=O)O
Melting Point 126.0°C
Packaging Glass Bottle
Color White
Infrared Spectrum Authentic
Quantity 10g
Water 0.5% max.
Assay Percent Range 99%
Linear Formula H2NC(=NH)SO2H
Beilstein 03, I, 36
Fieser 04,506; 05,668; 06,586
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: insoluble in organic solvents
Formula Weight 108.11
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Formamidinesulfinic acid, 99%
Show More Show Less

GHS H Statement
Self-heating in large quantities; may catch fire.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
May cause damage t

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep cool.
Protect from sunlight.
Avoid breathing dust/fume/ga

GHS Signal Word: Danger

EINECSNumber : 217-157-8

TSCA : TSCA