Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 88%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle

Chemical Identifiers

64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
CHEBI:30751
formic acid
C(=O)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 46.03
Packaging Glass Bottle
Specific Gravity 1L = 1.20kg
Quantity 2.5L
Flash Point 50.00°C
Chemical Name or Material Formic Acid
Assay 88%
Grade Baker Analyzed™ ACS Reagent