Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 90%, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 46.03
Percent Purity 90%
Grade BAKER ANALYZED™ Reagent
Packaging Glass Bottle
Specific Gravity 1L = 1.20kg
Quantity 500mL
Flash Point 50.00°C
Chemical Name or Material Formic Acid, Polymer Characterization Solvent