Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 96%, Reagent, ACS, 96%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
64-18-6
CH2O2
46.03
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
OC=O
64-18-6
CH2O2
46.03
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
OC=O

F1089, 64-18-6, CH2O2

Spectrum™ Formic Acid, 96 Percent, Reagent, ACS , also known as methanoic acid, occurs naturally in ant venom. One of its biggest uses is as a preservative agent in livestock feed. It is equally used in the production of leather goods. In the lab it is used as a source of formyl or hydride ion. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
CAS Max % 1
UN Number UN1779
Percent Purity 96%
Packaging Amber Glass Bottle
Quantity 6 x 500 mL