Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic acid, 98+%, pure, ACROS Organics™

Click to view available options
Quantity:
25mL
250mL
1L
2.5L
10L
Packaging:
Plastic bottle
Plastic drum
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
Additional Information Vapor Pressure: 44mbar at 20°C
Assay Percent Range 98+%
Linear Formula HCO2H
Beilstein 02, 8
Fieser 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
Merck Index 15, 4269
Density 1.2200g/mL
Formula Weight 46.02
Physical Form Liquid
Percent Purity ≥98.0%
Grade Pure
Infrared Spectrum Authentic
Packaging Plastic bottle
Refractive Index 1.3701 to 1.3721 (20°C, 589nm)
Color Undesignated
Quantity 25mL
Chemical Name or Material Formic acid
Show More Show Less