Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, ACS, 88%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
64-18-6,7732-18-5
64-18-6,7732-18-5
CAS Max % 1
Ammonium 5e-05
Chloride 1e-05
Dilution Test To Pass Test
Percent Purity 0.88
Grade ACS
Heavy Metals 5 ppm
Iron (Fe) 5 ppm
Packaging Glass Bottle
Residue after Evaporation 2e-05
Sulfate 2e-05
Quantity 2.5 L
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.