Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Formic acid, ACS, 96+%

Manufacturer:  Alfa Aesar™ 3661757

Quantity 4 x 500g

 View more versions of this product

Catalog No. AA3661757


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

64-18-6
HCOOH
MFCD00003297
1209246
Formic acid, For analysis ACS
CH2O2
46.03
UN1779
14,4241
ACS Grade

Specifications

64-18-6
>96+%
HCOOH
MFCD00003297
1209246
1.220g/mL
7° to 9°C
100° to 101°C
50° (156°F)
≥96%
Liquid
CH2O2
46.03
UN1779
14,4241
1.3710
Colorless
4 x 500g
Formic acid, For analysis ACS
ACS Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H314-H318-H226-H302
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FORMIC ACID

RTECSNumber : LQ4900000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit