Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid C-d 98atom%D, TCI America™

Click to view available options
Quantity:
1g
5g
917-71-5
CH2O2
47.031
MFCD00059016
BDAGIHXWWSANSR-MICDWDOJSA-N
11263466
deuterioformic acid
C(=O)O
917-71-5
CH2O2
47.031
MFCD00059016
BDAGIHXWWSANSR-MICDWDOJSA-N
11263466
deuterioformic acid
C(=O)O
Color Colorless
Quantity 1g
UN Number 1779
Formula Weight 47.03
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material Formic Acid C-d 98atom%D

EINECSNumber : (2)-0670

TSCA : No