Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, FCC, 85%, Spectrum™

Click to view available options
Quantity:
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
64-18-6
CH2O2
46.03
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
OC=O
64-18-6
CH2O2
46.03
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
OC=O

F1135, 64-18-6, CH2O2

Spectrum™ Formic Acid, FCC is the simplest carboxylic acid and one of its biggest uses is as a preservative and antibacterial agent in livestock feed. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
UN Number UN1779
Percent Purity 85%
Packaging Glass Bottle
Quantity 4 x 2.5 L