Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid Hydrazide 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
624-84-0
CH4N2O
60.056
XZBIXDPGRMLSTC-UHFFFAOYSA-N
formylhydrazine,formic acid hydrazide,formic hydrazide,hydrazinecarboxaldehyde,formhydrazide,n-formylhydrazine,carbazaldehyde,formylhydrazide,hydrazine, formyl,formal hydrazine
12229
formohydrazide
C(=O)NN
624-84-0
CH4N2O
60.056
XZBIXDPGRMLSTC-UHFFFAOYSA-N
formylhydrazine,formic acid hydrazide,formic hydrazide,hydrazinecarboxaldehyde,formhydrazide,n-formylhydrazine,carbazaldehyde,formylhydrazide,hydrazine, formyl,formal hydrazine
12229
formohydrazide
C(=O)NN
Color White
Infrared Spectrum Authentic
Quantity 25g
Melting Point 54°C to 58°C
Linear Formula HCONHNH2
Beilstein 02, 93
Formula Weight 60.06
Physical Form Shiny Crystalline Flakes
Percent Purity ≥97.5%
Chemical Name or Material Formic acid hydrazide, 98%

HYGROSCOPIC

EINECSNumber : 210-867-9

RTECSNumber : LQ8615000

TSCA : TSCA