Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProteoSpec™ Formic Acid, LCMS Grade, Ricca Chemical
SDP

Catalog Number rmb11202101
Click to view available options
Quantity:
10 x 1 mL
Packaging:
Glass Ampule
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O

LCMS Mobile Phase

pH Acidic
Melting Point 8.4°C
Boiling Point 100.8°C
CAS Min % 100
Color Colorless
CAS Max % 100
Solubility Information Miscible in water
Formula Weight 46.0254
Grade LC-MS
Density 1.2365
Quantity 10 x 1 mL
Packaging Glass Ampule
Physical Form Liquid
Chemical Name or Material ProteoSpec™ Formic Acid, LCMS Grade
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit