Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical

Click to view available options
Quantity:
0.5mL
1mL
2mL
50mL
10 x 1mL
Packaging:
Ampule
Ampules
Nalgene™ Poly Bottle
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O

Suitable as a mobile phase additive for LC/MS and UHPLC/MS

Absorbance 0.01 AU max. at 254nm,0.55 AU max. at 230nm,0.85 AU max. at 220nm,1.25 AU max. at 210nm
Additional Information Vapor Pressure: 44mbar at 20°C
Aluminum (Al) 20 ppb max.
Assay Percent Range ≥99.0 %
Calcium (Ca) 50 ppb max.
Linear Formula HCOOH
Chloride 5ppm max.
Merck Index 15, 4269
Density 1.220g/cm³
Copper (Cu) 10 ppb max.
Formula Weight 46.02
Physical Form Liquid
Percent Purity ≥99.0%
Grade OPTIMA™ LC-MS
Identification Pass Test
Iron (Fe) 10 ppb max.
Lead (Pb) 10 ppb max.
LCMS Gradient Suitability Pass Test
Manganese (Mn) 10 ppb max.
Magnesium (Mg) 10 ppb max.
Nickel (Ni) 10 ppb max.
Packaging Ampule
Potassium (K) 20 ppb max.
Residue after Evaporation 1ppm max.
Silver (Ag) 10 ppb max.
Sodium (Na) 50 ppb max.
Sulfate 50ppm max.
Sulfite 20ppm max.
Vapor Pressure 44mbar at 20°C
Zinc (Zn) 20 ppb max.
Melting Point 8°C
pH 2.1
Color Colorless
Boiling Point 101°C
Quantity 1mL
Chemical Name or Material Formic Acid
Show More Show Less

DANGER!

Emergency Overview
Flammable liquid and vapor. Causes severe burns by all exposure routes. Hygroscopic. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:2
Instability:0

DOTInformation : DOT Class 8, : Corrosive